2-Amino-5-bromo-4-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-4-(trifluoromethyl)pyridin-2-amine
PC48294-1G PC48294-5G PC48294-25G
APOSPC48294-1GEA 21.9 EUR
APOSPC48294-1G APOSPC48294-5G APOSPC48294-25G
2-Amino-5-bromo-4-(trifluoromethyl)pyridine
2-Amino-5-bromo-4-(trifluoromethyl)pyridine
Formula: C₆H₄BrF₃N₂
MW: 241.011 g/mol
Melting Pt: 66…75 °C
Storage Temperature: Ambient
MDL Number: MFCD09864896
CAS Number: 944401-56-3