α,α,α-Trifluoro-o-cresol 99%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Hydroxybenzotrifluoride, 2-(Trifluoromethyl)phenol
PC4830-100G PC4830-250G PC4830-25G
APOSPC4830-100GEA 60.9 EUR
APOSPC4830-100G APOSPC4830-250G APOSPC4830-25G
α,α,α-Trifluoro-o-cresol 99%
α,α,α-Trifluoro-o-cresol
Formula: C₇H₅F₃O
Boiling Pt: 147…148 °C
Melting Pt: 45…46 °C
Density: 1.3 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00002222
CAS Number: 444-30-4
EINECS: 207-148-7
UN: 1325
ADR: 4.1,II