4-Bromo-1-(difluoromethoxy)-2-fluorobenzene
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Difluoromethoxy)-3-fluorobromobenzene, 1-Bromo-3-fluoro-4-(difluoromethoxy)benzene
PC48300-1G PC48300-5G PC48300-25G
APOSPC48300-1GEA 30.6 EUR
APOSPC48300-1G APOSPC48300-5G APOSPC48300-25G
4-Bromo-1-(difluoromethoxy)-2-fluorobenzene
4-Bromo-1-(difluoromethoxy)-2-fluorobenzene
Formula: C₇H₄BrF₃O
MW: 241.0053 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11036067
CAS Number: 147992-27-6