2-Fluoro-4-iodo-5-methylpyridine
Supplier: Apollo Scientific
Danger
Synonyms: 2-Fluoro-4-iodo-5-picoline
PC48305-1G PC48305-25G PC48305-5G
APOSPC48305-1GEA 105 EUR
APOSPC48305-1G APOSPC48305-25G APOSPC48305-5G
2-Fluoro-4-iodo-5-methylpyridine
2-Fluoro-4-iodo-5-methylpyridine
Formula: C₆H₅FIN
MW: 237.02 g/mol
Melting Pt: 40…45 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095297
CAS Number: 153034-94-7