(1-Bromo-2,2,2-trifluoroethyl)benzene 95%
Supplier: Apollo Scientific
Warning
PC48327-5G PC48327-1G
APOSPC48327-5GEA 575 EUR
APOSPC48327-5G APOSPC48327-1G
(1-Bromo-2,2,2-trifluoroethyl)benzene 95%
(1-Bromo-2,2,2-trifluoroethyl)benzene
Formula: C₈H₆BrF₃
Boiling Pt: 49 °C/5mmHg
Density: 1.555 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD12789673
CAS Number: 434-42-4