2,3-Dibromo-5-fluorobenzotrifluoride 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1,2-Dibromo-5-fluoro-3-(trifluoromethyl)benzene
PC48350-10G PC48350-1G PC48350-5G
APOSPC48350-10GEA 389 EUR
APOSPC48350-10G APOSPC48350-1G APOSPC48350-5G
2,3-Dibromo-5-fluorobenzotrifluoride 97%
2,3-Dibromo-5-fluorobenzotrifluoride
Formula: C₇H₂Br₂F₄
MW: 321.89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800679
CAS Number: 1027511-90-5