α,α,α-Trifluoro-m-cresol 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3-(Trifluoromethyl)phenol, 3-Hydroxybenzotrifluoride
PC4840-25G PC4840-100G PC4840-500G
APOSPC4840-25GEA 21.9 EUR
APOSPC4840-25G APOSPC4840-100G APOSPC4840-500G
α,α,α-Trifluoro-m-cresol 98%
α,α,α-Trifluoro-m-cresol
Formula: C₇H₅F₃O
Boiling Pt: 178 °C
Melting Pt: –2 °C
Density: 1.333 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00002299
CAS Number: 98-17-9
EINECS: 202-645-5
UN: 2927
ADR: 6.1+8,II