4-Chloro-2-trifluoromethylpyrimidine 97%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Chloro-2-(trifluoromethyl)pyrimidine

PC48400-1G PC48400-25G PC48400-5G
APOSPC48400-1GEA 125 EUR
APOSPC48400-1G APOSPC48400-25G APOSPC48400-5G
4-Chloro-2-trifluoromethylpyrimidine 97%
4-Chloro-2-trifluoromethylpyrimidine
Formula: C₅H₂ClF₃N₂
MW: 182.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06411632
CAS Number: 1514-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR