5-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-oxazole-4-carboxylic acid
Supplier: Apollo Scientific
Warning
PC48401-1G PC48401-250MG PC48401-500MG
APOSPC48401-1GEA 478 EUR
APOSPC48401-1G APOSPC48401-250MG APOSPC48401-500MG
5-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-oxazole-4-carboxylic acid
5-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-oxazole-4-carboxylic acid
Formula: C₁₂H₈F₃NO₃
MW: 271.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07379226
CAS Number: 355020-55-2