3-Bromo-4-(trifluoromethoxy)benzonitrile
Supplier: Apollo Scientific
Warning
PC48427-1G PC48427-5G
APOSPC48427-1GEA 219 EUR
APOSPC48427-1G APOSPC48427-5G
3-Bromo-4-(trifluoromethoxy)benzonitrile
3-Bromo-4-(trifluoromethoxy)benzonitrile
Formula: C₈H₃BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD13195683
CAS Number: 191602-89-8