1-Bromo-3,5-difluoro-2-nitrobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-4,6-difluoronitrobenzene
PC48460-5G PC48460-100G PC48460-25G
APOSPC48460-5GEA 23.2 EUR
APOSPC48460-5G APOSPC48460-100G APOSPC48460-25G
1-Bromo-3,5-difluoro-2-nitrobenzene 98%
1-Bromo-3,5-difluoro-2-nitrobenzene
Formula: C₆H₂BrF₂NO₂
MW: 237.9900 g/mol
Melting Pt: 51…53 °C
Storage Temperature: Ambient
MDL Number: MFCD04112501
CAS Number: 884494-38-6