Perfluoroheptane (mixture of isomers) 98%
Supplier: Apollo Scientific
Warning
Synonyms: Hexadecafluoroheptane
PC48475-100G PC48475-25G PC48475-5G
APOSPC48475-100GEA 234 EUR
APOSPC48475-100G APOSPC48475-25G APOSPC48475-5G
Perfluoroheptane (mixture of isomers) 98%
Perfluoroheptane (mixture of isomers)
Formula: C₇F₁₆
MW: 388.05 g/mol
Boiling Pt: 80…82 °C
Density: 1.72 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040339
CAS Number: 335-57-9