2-(Methylsulphanyl)-6-(trifluoromethyl)-4-pyrimidinol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Methylthio)-6-(trifluoromethyl)pyrimidin-4-ol, 4-Hydroxy-2-(methylthio)-6-(trifluoromethyl)pyrimidine, 2-(Methylthio)-6-(trifluoromethyl)pyrimidin-4(1H)-one
PC4857-1G PC4857-5G PC4857-25G
APOSPC4857-1GEA 21.9 EUR
APOSPC4857-1G APOSPC4857-5G APOSPC4857-25G
2-(Methylsulphanyl)-6-(trifluoromethyl)-4-pyrimidinol 97%
2-(Methylsulphanyl)-6-(trifluoromethyl)-4-pyrimidinol
Formula: C₆H₅F₃N₂OS
MW: 210.18 g/mol
Melting Pt: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00153191
CAS Number: 16097-62-4