1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3(2H)-one
PC4858-25G PC4858-5G
APOSPC4858-25GEA 590 EUR
APOSPC4858-25G APOSPC4858-5G
1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol 97%
1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol
Formula: C₅H₅F₃N₂O
MW: 166.1 g/mol
Melting Pt: 129…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00085094
CAS Number: 119022-51-4