4-Hydroxy-8-methyl-2-(trifluoromethyl)quinoline
Supplier: Apollo Scientific
Warning
PC4858J-1G PC4858J-10G PC4858J-5G
APOSPC4858J-1GEA 58 EUR
APOSPC4858J-1G APOSPC4858J-10G APOSPC4858J-5G
4-Hydroxy-8-methyl-2-(trifluoromethyl)quinoline
4-Hydroxy-8-methyl-2-(trifluoromethyl)quinoline
Formula: C₁₁H₈F₃NO
Melting Pt: 125…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00276592
CAS Number: 1701-19-5