5-Nitro-3-(trifluoromethyl)pyridin-2-ol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Hydroxy-5-nitro-3-(trifluoromethyl)pyridine
PC4862-1G PC4862-250MG
APOSPC4862-1GEA 36.4 EUR
APOSPC4862-1G APOSPC4862-250MG
5-Nitro-3-(trifluoromethyl)pyridin-2-ol 98%
5-Nitro-3-(trifluoromethyl)pyridin-2-ol
Formula: C6H3F3N2O3
MW: 208.0948 g/mol
Melting Pt: 158 °C
Storage Temperature: Ambient
MDL Number: MFCD00276983
CAS Number: 99368-66-8