6-(Trifluoromethoxy)-4-quinolinol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Hydroxy-6-(trifluoromethoxy)quinoline, 6-(Trifluoromethoxy)quinolin-4-ol
PC4867-1G PC4867-250MG
APOSPC4867-1GEA 29 EUR
APOSPC4867-1G APOSPC4867-250MG
6-(Trifluoromethoxy)-4-quinolinol 97%
6-(Trifluoromethoxy)-4-quinolinol
Formula: C₁₀H₆F₃NO₂
MW: 229.16 g/mol
Melting Pt: 233…236 °C
Storage Temperature: Ambient
MDL Number: MFCD00219858
CAS Number: 175203-87-9