3-(Difluoromethoxy)benzonitrile 98%
Supplier: Apollo Scientific
Warning
PC4879-1G PC4879-5G PC4879-25G
APOSPC4879-1GEA 21.9 EUR
APOSPC4879-1G APOSPC4879-5G APOSPC4879-25G
3-(Difluoromethoxy)benzonitrile 98%
3-(Difluoromethoxy)benzonitrile
Formula: C₈H₅F₂NO
Boiling Pt: 224…225 °C
Density: 1.26 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236225
CAS Number: 97582-88-2
UN: 3439
ADR: 6.1,III