α,α,α-Trifluoro-2-iodotoluene 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Iodobenzotrifluoride, α,α,α-Trifluoro-o-iodotoluene, 1-Iodo-2-(trifluoromethyl)benzene
PC4890-10G
PC4890-100G
PC4890-500G
APOSPC4890-10GEA
38
EUR
APOSPC4890-10G
APOSPC4890-100G
APOSPC4890-500G
α,α,α-Trifluoro-2-iodotoluene 98%
α,α,α-Trifluoro-2-iodotoluene
Formula:
C₇H₄F₃I Boiling Pt: 197…198 °C Density: 1.936 g/cm³ (25 °C) Flash Pt: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001041 CAS Number: 444-29-1 EINECS: 207-147-1 UN: 3265 ADR: 8,II |