α,α,α-Trifluoro-3-iodotoluene 97%
Supplier: Apollo Scientific
Danger
Synonyms: α,α,α-Trifluoro-m-iodotoluene, 3-Iodobenzotrifluoride, 1-Iodo-3-(trifluoromethyl)benzene
PC4891-100G PC4891-1KG PC4891-500G
APOSPC4891-100GEA 125 EUR
APOSPC4891-100G APOSPC4891-1KG APOSPC4891-500G
α,α,α-Trifluoro-3-iodotoluene 97%
α,α,α-Trifluoro-3-iodotoluene
Formula: C₇H₄F₃I
MW: 272.01 g/mol
Boiling Pt: 82 °C/25mmHg
Melting Pt: –8 °C
Density: 1.887 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00001049
CAS Number: 401-81-0
EINECS: 206-934-7
UN: 3265
ADR: 8,II