α,α,α-Trifluoro-4-iodotoluene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Iodobenzotrifluoride, 1-Iodo-4-(trifluoromethyl)benzene, α,α,α-Trifluoro-p-iodotoluene
PC4892-100G PC4892-25G
APOSPC4892-100GEA 65.4 EUR
APOSPC4892-100G APOSPC4892-25G
α,α,α-Trifluoro-4-iodotoluene 98%
α,α,α-Trifluoro-4-iodotoluene
Formula: C₇H₄F₃I
MW: 272.01 g/mol
Boiling Pt: 185…186 °C/745mmHg
Melting Pt: 17…17.5 °C
Density: 1.851 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00039398
CAS Number: 455-13-0
EINECS: 207-234-4