6-Fluoro-1H-indazole 95%
Supplier: Apollo Scientific
Warning
PC49002-5G PC49002-1G
APOSPC49002-5GEA 89.9 EUR
APOSPC49002-5G APOSPC49002-1G
6-Fluoro-1H-indazole 95%
6-Fluoro-1H-indazole
Formula: C₇H₅FN₂
MW: 136.1264 g/mol
Melting Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD07371562
CAS Number: 348-25-4