4-Chloro-8-methyl-2-(trifluoromethyl)quinoline
Supplier: Apollo Scientific
PC49049-1G PC49049-5G
APOSPC49049-1GEA 128 EUR
APOSPC49049-1G APOSPC49049-5G
4-Chloro-8-methyl-2-(trifluoromethyl)quinoline
4-Chloro-8-methyl-2-(trifluoromethyl)quinoline
Formula: C₁₁H₇ClF₃N
Boiling Pt: 60 °C/0.2mmHg
Melting Pt: 22…24 °C
Storage Temperature: Ambient
MDL Number: MFCD08448258
CAS Number: 1701-25-3