2'-Methoxy-4'-(trifluoromethoxy)acetophenone 98%
Supplier: Apollo Scientific
Warning
PC49066-10G PC49066-5G PC49066-1G
APOSPC49066-10GEA 412 EUR
APOSPC49066-10G APOSPC49066-5G APOSPC49066-1G
2'-Methoxy-4'-(trifluoromethoxy)acetophenone 98%
2'-Methoxy-4'-(trifluoromethoxy)acetophenone
Formula: C₁₀H₉F₃O₃
MW: 234.18 g/mol
Melting Pt: 53…55 °C
Storage Temperature: Ambient
MDL Number: MFCD04115964
CAS Number: 886500-08-9