3-Amino-5-(trifluoromethyl)benzonitrile 98%
Supplier: Apollo Scientific
Warning
PC49080-1G PC49080-5G PC49080-250MG
APOSPC49080-1GEA 27.7 EUR
APOSPC49080-1G APOSPC49080-5G APOSPC49080-250MG
3-Amino-5-(trifluoromethyl)benzonitrile 98%
3-Amino-5-(trifluoromethyl)benzonitrile
Formula: C8H5F3N2
Storage Temperature: Ambient
MDL Number: MFCD09955455
CAS Number: 49674-28-4