2-Fluoro-5-hydroxybenzaldehyde
Supplier: Apollo Scientific
Warning
PC49096-1G PC49096-5G PC49096-25G
APOSPC49096-1GEA 45.1 EUR
APOSPC49096-1G APOSPC49096-5G APOSPC49096-25G
2-Fluoro-5-hydroxybenzaldehyde
2-Fluoro-5-hydroxybenzaldehyde
Formula: C₇H₅FO₂
MW: 140.1100 g/mol
Melting Pt: 74…75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09038469
CAS Number: 103438-84-2