p-Tolylsulphur pentafluoride
Supplier: Apollo Scientific
Danger
Synonyms: 4-Methylphenylsulphur Pentafluoride
PC49104-1G PC49104-5G PC49104-250MG
APOSPC49104-1GEA 284 EUR
APOSPC49104-1G APOSPC49104-5G APOSPC49104-250MG
p-Tolylsulphur pentafluoride
p-Tolylsulphur pentafluoride
Formula: C₇H₇F₅S
Boiling Pt: 80…81 °C/54mmHg
Density: 1.414 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16817600
CAS Number: 203126-21-0
UN: 2810
ADR: 6.1,III