2-Fluorophenylsulphur pentafluoride 97%
Supplier: Apollo Scientific
Warning
PC49115-1G PC49115-250MG
APOSPC49115-1GEA 178 EUR
APOSPC49115-1G APOSPC49115-250MG
2-Fluorophenylsulphur pentafluoride 97%
2-Fluorophenylsulphur pentafluoride
Formula: C₆H₄F₆S
Boiling Pt: 153…155 °C/637mmHg
Storage Temperature: Ambient
MDL Number: MFCD18909664
CAS Number: 864230-02-4
UN: 1993
ADR: 3,III