4-[(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)amino]-4-oxo-2-butenoic acid
Supplier: Apollo Scientific
PC4914-5G PC4914-500MG PC4914-1G
APOSPC4914-5GEA 1510 EUR
APOSPC4914-5G APOSPC4914-500MG APOSPC4914-1G
4-[(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)amino]-4-oxo-2-butenoic acid
4-[(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)amino]-4-oxo-2-butenoic acid
Formula: C₁₂H₁₁ClF₃N₃O₃
MW: 337.69 g/mol
Melting Pt: 166…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00202686
CAS Number: 339096-60-5