5-Bromo-2-(difluoromethyl)pyridine 98%
Supplier: Apollo Scientific
Danger
PC49144-1G PC49144-5G
APOSPC49144-1GEA 58 EUR
APOSPC49144-1G APOSPC49144-5G
5-Bromo-2-(difluoromethyl)pyridine 98%
5-Bromo-2-(difluoromethyl)pyridine
Formula: C₆H₄BrF₂N
Melting Pt: 35…40 °C
Flash Pt: 89.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11977429
CAS Number: 845827-13-6
UN: 2811
ADR: 6.1,III