3-Bromo-5-fluoro-2-methoxypyridine 98%
Supplier: Apollo Scientific
Warning
PC49160-25G
APOSPC49160-25GEA 81.2 EUR
APOSPC49160-25G
3-Bromo-5-fluoro-2-methoxypyridine 98%
3-Bromo-5-fluoro-2-methoxypyridine
Formula: C₆H₅BrFNO
Storage Temperature: Refrigerator
MDL Number: MFCD04972397
CAS Number: 884494-81-9