(4-(Trifluoromethyl)phenyl)methanesulphonyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: [4-(Trifluoromethyl)phenyl]methanesulphonyl chloride, 4-Trifluoromethyl-α-toluenesulphonyl chloride
PC49173-1G PC49173-250MG
APOSPC49173-1GEA 51 EUR
APOSPC49173-1G APOSPC49173-250MG
(4-(Trifluoromethyl)phenyl)methanesulphonyl chloride
(4-(Trifluoromethyl)phenyl)methanesulphonyl chloride
Formula: C₈H₆ClF₃O₂S
MW: 258.65 g/mol
Melting Pt: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD01631932
CAS Number: 163295-75-8
UN: 3261
ADR: 8 II