Difluoroiodomethane 10% (w/w) in THF
Supplier: Apollo Scientific
Danger
PC49177-10G PC49177-25G
APOSPC49177-10GEA 255 EUR
APOSPC49177-10G APOSPC49177-25G
Difluoroiodomethane 10% (w/w) in THF
Difluoroiodomethane
Formula: CHF₂I
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD03094113
CAS Number: 1493-03-4
UN: 2056
ADR: 3,II