2-Iodoperfluoropropane 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,1,1,2,3,3,3-Heptafluoro-2-iodopropane, Heptafluoroisopropyl iodide, Heptafluoro-2-iodopropane, 2-Iodoheptafluoropropane
PC4920-100G PC4920-10G PC4920-25G PC4920-500G
APOSPC4920-100GEA 78.3 EUR
APOSPC4920-100G APOSPC4920-10G APOSPC4920-25G APOSPC4920-500G
2-Iodoperfluoropropane 98%
2-Iodoperfluoropropane
Formula: C₃F₇I
MW: 295.93 g/mol
Boiling Pt: 38 °C
Melting Pt: –56 °C
Density: 2.1 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00042089
CAS Number: 677-69-0
EINECS: 211-643-3