2-Bromo-1-fluoro-4-methoxybenzene
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-4-fluoroanisole
PC49237-1G PC49237-25G PC49237-5G
APOSPC49237-1GEA 21.9 EUR
APOSPC49237-1G APOSPC49237-25G APOSPC49237-5G
2-Bromo-1-fluoro-4-methoxybenzene
2-Bromo-1-fluoro-4-methoxybenzene
Formula: C₇H₆BrFO
MW: 205.0200 g/mol
Boiling Pt: 81…82 °C/8mmHg
Storage Temperature: Ambient
MDL Number: MFCD13185359
CAS Number: 1161497-23-9