2-Methyl-3-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC49261-10G PC49261-1G PC49261-5G PC49261-25G
APOSPC49261-10GEA 292 EUR
APOSPC49261-10G APOSPC49261-1G APOSPC49261-5G APOSPC49261-25G
2-Methyl-3-(trifluoromethyl)benzaldehyde 98%
2-Methyl-3-(trifluoromethyl)benzaldehyde
Formula: C₉H₇F₃O
MW: 188.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116032
CAS Number: 878001-20-8