2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(3,4-dimethoxyphenyl)acetonitrile 97%
Supplier: Apollo Scientific
PC4927-1G PC4927-500MG PC4927-5G
APOSPC4927-1GEA 275 EUR
APOSPC4927-1G APOSPC4927-500MG APOSPC4927-5G
2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(3,4-dimethoxyphenyl)acetonitrile 97%
2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(3,4-dimethoxyphenyl)acetonitrile
Formula: C₁₆H₁₂ClF₃N₂O₂
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00172216
CAS Number: 338407-18-4