5-Fluoro-o-toluidine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Amino-4-fluorotoluene, 5-Fluoro-2-methylaniline
PC49276-100G PC49276-1KG PC49276-500G
APOSPC49276-100GEA 39.3 EUR
APOSPC49276-100G APOSPC49276-1KG APOSPC49276-500G
5-Fluoro-o-toluidine 97%
5-Fluoro-o-toluidine
Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 98…100 °C/15mmHg
Melting Pt: 37…40 °C
Density: 1.13
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00007764
CAS Number: 367-29-3
EINECS: 206-689-6
UN: 2811
ADR: 6.1 III