3'-(Trifluoromethyl)acetophenone 96%
Supplier: Apollo Scientific
Warning
Synonyms: m-(Trifluoromethyl)acetophenone, 1-[3-(Trifluoromethyl)phenyl]ethan-1-one
PC49336-25G PC49336-500G
APOSPC49336-25GEA 0 EUR
APOSPC49336-25G APOSPC49336-500G
3'-(Trifluoromethyl)acetophenone 96%
3'-(Trifluoromethyl)acetophenone
Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 198…200 °C
Density: 1.235 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000391
CAS Number: 349-76-8