1-Iodo-3,3,3-trifluoropropane 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,1,1-Trifluoro-3-iodopropane, 3-Iodo-1,1,1-trifluoropropane
PC4946-25G PC4946-5G
APOSPC4946-25GEA 43.5 EUR
APOSPC4946-25G APOSPC4946-5G
1-Iodo-3,3,3-trifluoropropane 98%
1-Iodo-3,3,3-trifluoropropane
Formula: C₃H₄F₃I
MW: 223.96 g/mol
Boiling Pt: 80 °C
Density: 1.911 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00038531
CAS Number: 460-37-7