2-Bromo-4-(trifluoromethyl)phenol
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-4-hydroxybenzotrifluoride
PC49531-5G PC49531-25G PC49531-100G
APOSPC49531-5GEA 21.9 EUR
APOSPC49531-5G APOSPC49531-25G APOSPC49531-100G
2-Bromo-4-(trifluoromethyl)phenol
2-Bromo-4-(trifluoromethyl)phenol
Formula: C₇H₄BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD06657954
CAS Number: 81107-97-3