Isopropyl 4,4,4-trifluoroacetoacetate 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4,4,4-Trifluoroacetoacetic Acid Isopropyl Ester
PC4956-25G
APOSPC4956-25GEA 61.1 EUR
APOSPC4956-25G
Isopropyl 4,4,4-trifluoroacetoacetate 97%
Isopropyl 4,4,4-trifluoroacetoacetate
Formula: C₇H₉F₃O₃
MW: 198.14 g/mol
Boiling Pt: 42 °C/28mmHg
Storage Temperature: Ambient
MDL Number: MFCD00040990
CAS Number: 175230-50-9