1-Chloro-2,6-difluorobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-1,3-difluorobenzene
PC4958-5G PC4958-1G PC4958-25G
APOSPC4958-5GEA 21.9 EUR
APOSPC4958-5G APOSPC4958-1G APOSPC4958-25G
1-Chloro-2,6-difluorobenzene 98%
1-Chloro-2,6-difluorobenzene
Formula: C₆H₃ClF₂
MW: 148.5378 g/mol
Boiling Pt: 131 °C
Density: 1.37 g/cm³ (25 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00142604
CAS Number: 38361-37-4
UN: 1993
ADR: 3,III