1-Bromo-3-fluoro-5-nitrobenzene
Supplier: Apollo Scientific
Warning
PC49594-25G PC49594-5G
APOSPC49594-25GEA 109 EUR
APOSPC49594-25G APOSPC49594-5G
1-Bromo-3-fluoro-5-nitrobenzene
1-Bromo-3-fluoro-5-nitrobenzene
Formula: C₆H₃BrFNO₂
MW: 220.00 g/mol
Boiling Pt: 65…70 °C
Melting Pt: 27…29 °C
Storage Temperature: Ambient
MDL Number: MFCD09955454
CAS Number: 7087-65-2