2-Chloro-6-(trifluoromethyl)benzoyl chloride 96%
Supplier: Apollo Scientific
Danger
PC49644-25G PC49644-5G PC49644-1G
APOSPC49644-25GEA 221 EUR
APOSPC49644-25G APOSPC49644-5G APOSPC49644-1G
2-Chloro-6-(trifluoromethyl)benzoyl chloride 96%
2-Chloro-6-(trifluoromethyl)benzoyl chloride
Formula: C₈H₃Cl₂F₃O
MW: 243.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09025361
CAS Number: 916420-44-5
UN: 3265
ADR: 8 II