2-Bromo-1-fluoro-3-iodobenzene, Technical Grade
Supplier: Apollo Scientific
Warning
PC49675-1G PC49675-5G
APOSPC49675-1GEA 0 EUR
APOSPC49675-1G APOSPC49675-5G
2-Bromo-1-fluoro-3-iodobenzene, Technical Grade
2-Bromo-1-fluoro-3-iodobenzene
Formula: C₆H₃BrFI
MW: 300.8949 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09056796
CAS Number: 851368-08-6