2-Amino-5-fluorobenzonitrile 99%
Supplier: Apollo Scientific
Warning
PC4978-25G PC4978-5G PC4978-100G
APOSPC4978-25GEA 69.6 EUR
APOSPC4978-25G APOSPC4978-5G APOSPC4978-100G
2-Amino-5-fluorobenzonitrile 99%
2-Amino-5-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.13 g/mol
Boiling Pt: 138 °C/15mmHg
Melting Pt: 92…96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683869
CAS Number: 61272-77-3