1-(2,6-Difluorophenyl)ethanol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,6-Difluoro-alpha-methylbenzyl alcohol
PC50008-25G
PC50008-5G
APOSPC50008-25GEA
344
EUR
APOSPC50008-25G
APOSPC50008-5G
1-(2,6-Difluorophenyl)ethanol
1-(2,6-Difluorophenyl)ethanol
Formula:
C₈H₈F₂O MW: 158.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00274321 CAS Number: 87327-65-9 |