N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-N-cyclopropylamine
Supplier: Apollo Scientific
Warning
Synonyms: 3-Chloro-2-(cyclopropylamino)-5-(trifluoromethyl)pyridine
PC50017-1G PC50017-5G
APOSPC50017-1GEA 174 EUR
APOSPC50017-1G APOSPC50017-5G
N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-N-cyclopropylamine
N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-N-cyclopropylamine
Formula: C₉H₈ClF₃N₂
MW: 236.63 g/mol
Melting Pt: 49…50 °C
Storage Temperature: Ambient
MDL Number: MFCD11117304
CAS Number: 1031619-96-1