2-Methyl-3-(trifluoromethyl)phenylacetonitrile 98%
Supplier: Apollo Scientific
Danger
PC500182-1G PC500182-5G
APOSPC500182-1GEA 136 EUR
APOSPC500182-1G APOSPC500182-5G
2-Methyl-3-(trifluoromethyl)phenylacetonitrile 98%
2-Methyl-3-(trifluoromethyl)phenylacetonitrile
Formula: C₁₀H₈F₃N
MW: 199.17 g/mol
Melting Pt: 51…55 °C
Storage Temperature: Ambient
MDL Number: MFCD09832278
CAS Number: 1000515-00-3
UN: 3439
ADR: 6.1 III